Buffer solution pH 10, Carbonate buffer
Buffer solution pH 10, Carbonate buffer
Thermo Scientific Chemicals

Buffer solution pH 10, Carbonate buffer

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Catalog number 258605000
Price (USD)
52.30
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
52.30
Each
Add to cart
Specifications
CAS1332-57-6
CAS144-55-8
CAS7732-18-5
Chemical Name or MaterialBuffer solution pH 10
ColorColorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH buffer
RUO – Research Use Only

Share catalog number, name or link