(Aminomethyl)cyclopropane, 97%, Thermo Scientific Chemicals
(Aminomethyl)cyclopropane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(Aminomethyl)cyclopropane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
10 g
Packaging:
Glass bottle
Catalog number 258720500
Price (USD)/ Each
873.00
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
873.00
Add to cart
(Aminomethyl)cyclopropane, 97%, Thermo Scientific Chemicals
Catalog number258720500
Price (USD)/ Each
873.00
-
Add to cart
Chemical Identifiers
CAS2516-47-4
IUPAC Namecyclopropylmethanaminium
Molecular FormulaC4H10N
InChI KeyIGSKHXTUVXSOMB-UHFFFAOYSA-O
SMILES[NH3+]CC1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.4300 to 1.4340 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only