1-Ethoxy-2-propanol, 95%, Thermo Scientific Chemicals
1-Ethoxy-2-propanol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Ethoxy-2-propanol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 259050025
Price (USD)/ Each
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1-ethoxypropan-2-ol
Molecular FormulaC5H12O2
InChI KeyJOLQKTGDSGKSKJ-UHFFFAOYNA-N
SMILESCCOCC(C)O
Molecular Weight (g/mol)104.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Water=<0.1 % (K.F.)
Appearance (Form)Liquid
GC>=94 % Remark: balance 2-ethoxy-1-propanol
Refractive index1.4050 to 1.4090 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only