Thermo Scientific Chemicals

Amidinothiourea, 99+%, Thermo Scientific Chemicals

Catalog number: 259190100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Amidinothiourea, 99+%, Thermo Scientific Chemicals

Catalog number: 259190100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 259190100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2114-02-5
IUPAC Name
(diaminomethylidene)thiourea
Molecular Formula
C2H6N4S
InChI Key
OKGXJRGLYVRVNE-UHFFFAOYSA-N
SMILES
NC(N)=NC(N)=S
Infrared spectrum
Conforms
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder or crystals
Titration Iodimetric
>=99.0 %
Loss on drying
=<0.3 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text