γ-Undecalactone, 98%, Thermo Scientific Chemicals
γ-Undecalactone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

γ-Undecalactone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
500 mL
Packaging:
Glass bottle
Catalog number 259491000
Price (USD)/ Each
47.40
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
47.40
Add to cart
γ-Undecalactone, 98%, Thermo Scientific Chemicals
Catalog number259491000
Price (USD)/ Each
47.40
-
Add to cart
Chemical Identifiers
CAS104-67-6
IUPAC Name(5S)-5-heptyloxolan-2-one
Molecular FormulaC11H20O2
InChI KeyPHXATPHONSXBIL-JTQLQIEISA-N
SMILESCCCCCCC[C@H]1CCC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4490 to 1.4540 (20°C, 589 nm)
Specific gravity(25°C) 0.939 to 0.948
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only