Thermo Scientific Chemicals

γ-Undecalactone, 98%, Thermo Scientific Chemicals

Catalog number: 259495000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

γ-Undecalactone, 98%, Thermo Scientific Chemicals

Catalog number: 259495000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 259495000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
104-67-6
IUPAC Name
(5S)-5-heptyloxolan-2-one
Molecular Formula
C11H20O2
InChI Key
PHXATPHONSXBIL-JTQLQIEISA-N
SMILES
CCCCCCC[C@H]1CCC(=O)O1
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4490 to 1.4540 (20°C, 589 nm)
Specific gravity
(25°C) 0.939 to 0.948

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text