Thermo Scientific Chemicals

Potassium tetraborate tetrahydrate, 99.5%, extra pure, Thermo Scientific Chemicals

Catalog number: 259570010
1 kg, Each
Thermo Scientific Chemicals

Potassium tetraborate tetrahydrate, 99.5%, extra pure, Thermo Scientific Chemicals

Catalog number: 259570010
1 kg, Each
Quantity
Catalog number: 259570010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12045-78-2
IUPAC Name
potassium tetrahydrate borate
Molecular Formula
BH8KO7
InChI Key
SAAWPUYPXWYTFF-UHFFFAOYSA-N
SMILES
O.O.O.O.[K+].[O-]B([O-])[O-]
Appearance (Form)
Powder
Iron (Fe)
=<20 ppm
Heavy metals (as Pb)
=<20 ppm
Chloride (Cl)
=<20 ppm
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text