Butyl butyrate, 98%, Thermo Scientific Chemicals
Butyl butyrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Butyl butyrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
Packaging:
Glass bottle
Catalog number 259590010
Price (USD)/ Each
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS109-21-7
IUPAC Namebutyl butanoate
Molecular FormulaC8H16O2
InChI KeyXUPYJHCZDLZNFP-UHFFFAOYSA-N
SMILESCCCCOC(=O)CCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Specific gravity(25°C) 0.866 to 0.869
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only