2-Phenylbenzimidazole-5-sulfonic acid, 98%, Thermo Scientific Chemicals
2-Phenylbenzimidazole-5-sulfonic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Phenylbenzimidazole-5-sulfonic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 259702500
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
187.00 
Save 28.35 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
187.00 
Save 28.35 (15%)
Add to cart
2-Phenylbenzimidazole-5-sulfonic acid, 98%, Thermo Scientific Chemicals
Catalog number259702500
Price (USD)/ Each
158.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
187.00 
Save 28.35 (15%)
-
Add to cart
Chemical Identifiers
CAS27503-81-7
IUPAC Name2-phenyl-1H-1,3-benzodiazole-6-sulfonic acid
Molecular FormulaC13H10N2O3S
InChI KeyUVCJGUGAGLDPAA-UHFFFAOYSA-N
SMILESOS(=O)(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration excess NaOH>=97.5 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Water=<7 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only