Thermo Scientific Chemicals

Isobutyl acetate, 98%, Thermo Scientific Chemicals

Catalog number: 259830025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Isobutyl acetate, 98%, Thermo Scientific Chemicals

Catalog number: 259830025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 259830025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
110-19-0
IUPAC Name
2-methylpropyl acetate
Molecular Formula
C6H12O2
InChI Key
GJRQTCIYDGXPES-UHFFFAOYSA-N
SMILES
CC(C)COC(C)=O
Infrared spectrum
Conforms
Appearance (Form)
Clear liquid
GC
>=97.5 %
Density
(20°C) 0.871 to 0.875 kg/l
Acidity (CH3COOH)
=<0.01 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text