Thermo Scientific Chemicals

Sodium hydroxide, extra pure, 50 wt% solution in water, Thermo Scientific Chemicals

Catalog number: 259860251
25 L, Plastic drum, Each
Thermo Scientific Chemicals

Sodium hydroxide, extra pure, 50 wt% solution in water, Thermo Scientific Chemicals

Catalog number: 259860251
25 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 259860251
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1310-73-2
IUPAC Name
sodium hydroxide
Molecular Formula
HNaO
InChI Key
HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES
[OH-].[Na+]
Appearance (Form)
Viscous solution
Sodium carbonate (Na2CO3)
=<0.2 % (Na2CO3)
Sodium sulfate (Na2SO4)
=<100 ppm (Na2SO4)
Appearance (Color)
Clear colorless
Titration with HCl
49.0 to 51.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text