TraceGOLD TG-POLAR GC Columns
TraceGOLD TG-POLAR GC Columns
Thermo Scientific™

TraceGOLD TG-POLAR GC Columns

Redefine your expectations for high-polarity GC columns with the Thermo Scientific TraceGold TG-POLAR Columns
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric) InnerFilm Thickness
26082-5030105 m0.25 mm0.2 μm
26082-047030 m0.25 mm0.1 μm
26082-059060 m0.25 mm0.1 μm
26082-5000105 m0.25 mm0.1 μm
26082-501030 m0.25 mm0.2 μm
26082-502060 m0.25 mm0.2 μm
26082-504030 m0.32 mm0.2 μm
26082-505060 m0.32 mm0.2 μm
26082-336030 m0.53 mm1.5 μm
Catalog number 26082-5030
Price (USD)
1,998.19
Each
Add to cart
Length (Metric):
105 m
Diameter (Metric) Inner:
0.25 mm
Film Thickness:
0.2 μm
Price (USD)
1,998.19
Each
Add to cart
Ask our AI about this Product

Refine your expectations for high-polarity GC/MS with the Thermo Scientific™ TraceGOLD™ TG-POLAR columns, which are specifically designed with a polymer and surface treatment to overcome traditional problems with high-polarity columns.

Engineered for Superior Performance

  • Highly polar phase, 90% biscyanopropyl/10% phenylcyanopropyl polysiloxane, not bonded
  • Strong dipole moment and high selectivity for cis/trans compounds or compounds with conjugated double bonds
  • Equivalent to U.S.P G8 and G48

Applications:

  • Cis/Trans fatty acid methyl esters (FAMEs)
  • Dioxins
A Leap Forward in Performance with Low Bleed and Superior Inertness:
  • A low bleed, stable column produces consistent results.
  • Low baseline noise enables improved limits of detection with enhanced resolution of low-level analytes.
  • Minimizing column bleed extends column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior intertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Alternative Uses:
  • Alcohols
  • Aldehydes
  • Biodiesel–FAMEs
Similar To:
  • DB-23
  • HP-23
  • Rtx™™-2330
  • SP™-2330
  • SP™-2380
  • SPB™-2560
  • HP-88
  • Silar™™ 90c
  • CP-Sil 88 FAME
  • CP-Sil 88
  • BPX 70
  • BPX 90
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description0.2 um Thickness; 0.25 mm ID; 105 m Length
Diameter (Metric)0.25 mm
For Use With (Application)Dioxins Analysis, GC/MS of Dioxins
Max. Temperature260/275°C
PolarityHighly Polar
Diameter (Metric) Inner0.25 mm
Film Thickness0.2 μm
Length (Metric)105 m
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-Polar
USP TypeG8, G48
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Videos

Video Player is loading.
Current Time 0:00
Duration 0:00
Loaded: 0%
Stream Type LIVE
Remaining Time 0:00
 
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Discover the capabilities of Trace GOLD GC columns, which offer a leap in column performance by offering ultra-low bleed, high levels of reproducibility and superior inertness, ensuring full confidence in your analysis.

      Product video duration: 2 minutes

      Documents & Downloads

      Certificates

      0 results displayed, search above for a specific certificate

      Share catalog number, name or link

      1x1 image pixel for data collection