96 Well Plate, Non-Treated, Non-Sterile, Pack of 10
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
96 Well Plate, Non-Treated, Non-Sterile, Pack of 10
Thermo Scientific™
96 Well Plate, Non-Treated, Non-Sterile, Pack of 10
Color:Clear; Sterility:Non-sterile; Lid:With Lid
Have Questions?
Catalog NumberQuantity
260895160/Cs.
Catalog number 260895
Price (USD)
762.00
Case of 160
-
Add to cart
Quantity:
160/Cs.
Price (USD)
762.00
Case of 160
Add to cart
Ask our AI about this Product
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Certifications/ComplianceAll microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Description96F well plate, Non-Treated, clear, with lid, Non-sterile (Pack of 10, Case of 160)
LidWith Lid
No. per Case160
No. per Pack10
Packaging10 Pk., 160 Case
Product TypeMicroplate
Quantity160/Cs.
Array8 x 12
BarcodeCustom Barcoding Available
ColorClear
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilityNon-sterile
Surface TreatmentNon-treated
Volume (Metric) Well400 μL
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 160
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link