Thermo Scientific™

MA(PEG)12 Methyl-PEG-Amine Compound

Catalog number: 26115
Have Questions?
Thermo Scientific™

MA(PEG)12 Methyl-PEG-Amine Compound

Catalog number: 26115
Have Questions?
Catalog Number
26115
Unit Size
1 g
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
261151 g
-
26114100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce MA(PEG)12 is a methyl- and amine-terminated polyethylene glycol reagent that is useful for a variety of surface-modification and molecule-pegylation applications.

Features of MA(PEG)12:

• Fully characterized PEGylation reagent with defined PEG chain length; discrete molecular weight for consistency of performance in protein-modification applications
• PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
• Allows site-specific labeling of primary amines or carboxyl groups on proteins or surfaces
• Easy-to-follow instructions increase the likelihood of a successful outcome

MA(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers and terminal methyl (-CH3) and amino (-NH2) groups. The unbranched, hydrophilic, discrete-length molecules have the form Methyl-PEGn-Amine, where the subscript 'n' denotes 4, 8, 12, or 24 ethylene glycol units. The terminal primary amine of each compound provides a specific target for crosslinking and other conjugation methods, making these compounds useful as PEGylation reagents.

Applications of PEGylation:
• PEGylate carboxylate or amine surfaces
• Add inert mass to proteins, immunogens, drug compounds and probes
• Improve solubility (decrease aggregation) of proteins or peptides without affecting function
• Protect proteins from proteolysis

Methods of Covalent Attachment:
• Conjugate to carboxyl groups using EDC and Sulfo-NHS
• Crosslink to primary amines using DSS, BS(PEG)5/BS(PEG)9 or other NHS-ester reagent
• Crosslink to sulfhydryl groups using Sulfo-SMCC or SM(PEG)n reagent
• Attach to oxidized carbohydrate groups (aldehydes) by reductive amination

Why PEGylate a protein or peptide?
Methyl-capped PEG-containing reagents have been used to modify proteins to provide specific advantages. Protein PEGylation can improve the stability of the modified protein, protect it from proteolytic digestion, increase its half life in a biological application, mask it from causing an immunogenic response, decrease its antigenicity or potential toxicity, improve its solubility, diminish the potential for aggregation, and minimize interference for both in vitro and in vivo applications. Polyethylene glycol, also called polyethylene oxide (PEO), has these effects because it is nontoxic, nonimmunogenic, hydrophilic, water soluble and highly flexible.

Advantages of Discrete-length Polyethylene Glycol Compounds:
These reagents are specially synthesized, resulting in homogeneous compounds of defined molecular weight, characterized by discrete chain lengths, providing a greater ability to optimize and characterize surface protein modifications. Typical preparations of PEG compounds are a heterogeneous mixtures composed of a distribution of chain lengths with a specified average molecular weight or approximate number of PEG subunits.

Related Products
MA(PEG)4 Methyl-PEG-Amine Compound
MA(PEG)8 Methyl-PEG-Amine Compound
MA(PEG)24 Methyl-PEG-Amine Compound
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Aldehyde
Description
Pierce MA(PEG)12
Label Type
Polyethylene Glycol (PEG), Blocking Groups
Reactive Moiety
Amine, Primary Amine
Spacer
12 ethylene glycol units
Form
Solid
Quantity
1 g
Solubility
Water
Product Type
Amine Compound

Contents & Storage

Upon receipt store product at -20°C protected from moisture. Product is shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text