Thermo Scientific™

CA(PEG)8 Carboxy-PEG-Amine Compound

Catalog number: 26123
Have Questions?
Thermo Scientific™

CA(PEG)8 Carboxy-PEG-Amine Compound

Catalog number: 26123
Have Questions?
Catalog Number
26123
also known as 26127
Unit Size
1 g
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
26123
also known as 26127
1 g
-
26122100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce CA(PEG)8 is a pegylated amino acid of the form carboxy-PEG-amine, containing 8 polyethylene glycol units, and is useful for a variety of surface- and molecule-pegylation applications.

CA(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers with carboxy (-COOH) and amino (-NH2) termini. The unbranched, hydrophilic, discrete-length molecules have the form Carboxy-PEGn-Amine, where the subscript 'n' denotes 4, 8, 12, or 24 ethylene glycol units. The carboxyl and primary amine of each compound provide specific targets for crosslinking and other conjugation methods, making these compounds useful as PEGylation reagents.

Features of CA(PEG)n Compounds

• Fully characterized PEGylation reagents with defined PEG chain lengths; molecules of discrete molecular weight for consistency of performance in protein-modification applications
• Provided as a series of 4, 8, 12 and 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application while retaining all the benefits associated with protein PEGylation
• PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
• Allows site-specific labeling or crosslinking involving primary amines or carboxyl groups on proteins or surfaces
• Easy-to-follow instructions increase the likelihood of a successful outcome

Applications of PEGylation
• PEGylate carboxyl or amine surfaces
• Add inert mass to proteins, immunogens, drug compounds and probes
• Improve solubility (decrease aggregation) of proteins or peptides without affecting function
• Protect proteins from proteolysis

Methods of Covalent Attachment
• Derivatize or crosslink carboxyls or primary amines using EDC and Sulfo-NHS
• Crosslink amino terminus to primary amines using DSS, BS(PEG)5/BS(PEG)9 or other NHS-ester reagent
• Crosslink amino terminus to sulfhydryl groups using Sulfo-SMCC or SM(PEG)n reagent

Why PEGylate a protein or peptide?
PEG-containing reagents have been used to modify proteins to provide specific advantages. Protein PEGylation can improve the stability of the modified protein, protect it from proteolytic digestion, increase its half life in biological applications, mask it from causing an immunogenic response, decrease its antigenicity or potential toxicity, improve its solubility, diminish the potential for aggregation, and minimize interference for both in vitro and in vivo applications. Polyethylene glycol, also called polyethylene oxide (PEO), has these effects because it is nontoxic, nonimmunogenic, hydrophilic, water soluble and highly flexible.

Advantages of Discrete-length Polyethylene Glycol Compounds
These reagents are specially synthesized, resulting in homogeneous compounds of defined molecular weight, characterized by discrete chain lengths, providing a greater ability to optimize and characterize surface protein modifications. Typical preparations of PEG compounds are heterogeneous mixtures composed of a distribution of chain lengths with a specified average molecular weight or approximate number of PEG subunits.

Related Products
CA(PEG)4 Carboxy-PEG-Amine Compound
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Carboxylic Acid
Description
Pierce CA(PEG)8
Label Type
Conjugating Groups, Polyethylene Glycol (PEG)
Reactive Moiety
Amine, Carboxylic Acid, Primary Amine
Spacer
8 ethylene glycol units
Form
Powder
Quantity
1 g
Solubility
Water
Product Type
Amine Compound

Contents & Storage

Store desiccated at 4°C. Shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text