1x1 image pixel for data collection
PEG4-SPDP (PEGylated, long-chain SPDP crosslinker)
PEG4-SPDP (PEGylated, long-chain SPDP crosslinker)
Thermo Scientific™
PEG4-SPDP (PEGylated, long-chain SPDP crosslinker)
Thermo Scientific Pierce PEG4-SPDP is a multifunctional crosslinker for protein conjugation via amine-to-amine or amine-to-sulfhydryl crosslinks that contain a 4-unitRead more
Have Questions?
Catalog NumberQuantity
26128100 mg
Catalog number 26128
Price (USD)
420.00
Each
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)
420.00
Each
Add to cart
Thermo Scientific Pierce PEG4-SPDP is a multifunctional crosslinker for protein conjugation via amine-to-amine or amine-to-sulfhydryl crosslinks that contain a 4-unit polyethylene glycol (PEG) group and a reducible (cleavable) disulfide bond.
Features of PEG4-SPDP:
Efficient amine conjugationNHS ester provides effective conjugation to lysines, N-termini of peptides and other primary amines
Reversible sulfhydryl conjugationpyridyldithiol group enables disulfide linkage with reduced cysteines and other sulfhydryl groups, providing a stable but cleavable bond
Polyethylene glycol—PEG groups are flexible, non-immunogenic, hydrophilic, and often enhance the solubility of attached molecules
Spacer arm—connects primary amines and sulfhydryl targets at distances up to 25.7 angstroms; compare to PEG12-SPDP (54.1 angstroms)
SPDP-type reagents have an amine-reactive N-hydroxysuccinimide (NHS) ester at one end and a suflhydryl-reactive 2-pyridyldithiol group at the opposite end. PEG4-SPDP has a 4-unit polyethylene glycol spacer arm, which confers greater solubility to the crosslinker and linked proteins compared to crosslinkers having only hydrocarbon spacers. Pyridyldithiol reagents produce disulfide-containing linkages that can be cleaved with reducing agents such as dithiothreitol (DTT). Crosslinking experiments with SPDP reagents are not limited to those involving proteins. Any of a variety of molecules with primary amines and sulfhydryl groups can be modified or crosslinked using an SPDP reagent.
Conjugation of SPDP reagents to primary amines reactions via the NHS ester is most commonly performed at pH 7 to 8 in phosphate, carbonate/bicarbonate or borate buffer. Other buffers that do not contain primary amines (or thiols or disulfide reducing reagents—see the sulfhydryl reactivity discussion) are also effective.
Reaction with the sulfhydryl-reactive pyridyldithio group can be performed with (simultaneously) or following the amine reaction, and it occurs optimally at pH 7 to 8. The reaction results in pyridine-2-thione displacement, the concentration of which can be determined by measuring the absorbance at 343nm. Reaction buffers must be free of thiols and disulfide reducing agents until quenching or reduction of the 2-pyridyldithio is desired.
Product References:
Crosslinker Application Guide -- search for recent literature references for this product
Related Products
PEG12-SPDP (PEGylated, long-chain SPDP crosslinker)
SPDP (succinimidyl 3-(2-pyridyldithio)propionate)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityNo
DescriptionPEG4-SPDP
FormLiquid
Labeling MethodChemical Labeling
Molecular Weight (g/mol)559.65
PEGylatedYes
Product LinePierce™
Quantity100 mg
Reactive MoietyNHS Ester, Pyridyldisulfide
Shipping ConditionAmbient
SolubilityWater
Spacer Arm Length25.7 Å
Water SolubleYes
Chemical ReactivityAmine-Sulfhydryl
CleavableBy Thiols
Crosslinker TypeHeterobifunctional
FormatStandard
Product TypeCrosslinker
SpacerMedium (10 to 30 Å)
Unit SizeEach
Contents & Storage
Upon receipt store at -20°C protected from moisture.
Frequently asked questions (FAQs)
Can you provide the shelf-life for PEG4-SPDP (PEGylated, long-chain SPDP crosslinker)?
PEG4-SPDP (PEGylated, long-chain SPDP crosslinker)Sulfo-SDAD (Sulfo-NHS-SS-Diazirine) (sulfosuccinimidyl 2-((4,4'-azipentanamido)ethyl)-1,3'-dithiopropionate) is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link