Thermo Scientific™

PEG12-SPDP (PEGylated, long-chain SPDP crosslinker)

Catalog number: 26129
Have Questions?
Thermo Scientific™

PEG12-SPDP (PEGylated, long-chain SPDP crosslinker)

Catalog number: 26129
Have Questions?
Catalog Number
26129
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
26129100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce PEG12-SPDP is a multifunctional crosslinker for protein conjugation via amine-to-amine or amine-to-sulfhydryl crosslinks that contain a 12-unit polyethylene glycol (PEG) group and a reducible (cleavable) disulfide bond.

Features of PEG12-SPDP:

Efficient amine conjugationNHS ester provides effective conjugation to lysines, N-termini of peptides and other primary amines
Reversible sulfhydryl conjugationpyridyldithiol group enables disulfide linkage with reduced cysteines and other sulfhydryl groups, providing a stable but cleavable bond
Polyethylene glycol—PEG groups are flexible, non-immunogenic, hydrophilic, and often enhance the solubility of attached molecules
Spacer arm—connects primary amines and sulfhydryl targets at distances up to 54.1 angstroms; compare to PEG4-SPDP (25.7 angstroms)

SPDP-type reagents have an amine-reactive N-hydroxysuccinimide (NHS) ester at one end and a suflhydryl-reactive 2-pyridyldithio group at the opposite end. PEG12-SPDP has a 12-unit polyethylene glycol spacer arm, which confers greater solubility to the crosslinker and linked proteins compared to crosslinkers having only hydrocarbon spacers. Pyridyldithiol reagents produce disulfide-containing linkages that can be cleaved with reducing agents such as dithiothreitol (DTT). Crosslinking experiments with SPDP reagents are not limited to those involving proteins. Any of a variety of molecules with primary amines and sulfhydryl groups can be modified or crosslinked using an SPDP reagent.

Conjugation of SPDP reagents to primary amines reactions via the NHS ester is most commonly performed at pH 7 to 8 in phosphate, carbonate/bicarbonate or borate buffer. Other buffers that do not contain primary amines (or thiols or disulfide reducing reagents—see the sulfhydryl reactivity discussion) are also effective.

Reaction with the sulfhydryl-reactive pyridyldithio group can be performed with (simultaneously) or following the amine reaction, and it occurs optimally at pH 7 to 8. The reaction results in pyridine-2-thione displacement, the concentration of which can be determined by measuring the absorbance at 343nm. Reaction buffers must be free of thiols and disulfide reducing agents until quenching or reduction of the 2-pyridyldithio is desired.

Product References:
Crosslinker Application Guide -- search for recent literature references for this product

Related Products
PEG4-SPDP (PEGylated, long-chain SPDP crosslinker)
SPDP (succinimidyl 3-(2-pyridyldithio)propionate)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Amine-Sulfhydryl
Cleavable
By Thiols
Description
PEG12-SPDP
Molecular Weight (g/mol)
912.07
PEGylated
Yes
Spacer Arm Length
54.1 Å
Cell Permeability
No
Shipping Condition
Ambient
Product Line
Pierce™
Labeling Method
Chemical Labeling
Crosslinker Type
Heterobifunctional
Reactive Moiety
NHS Ester, Pyridyldisulfide
Spacer
Long (>30 Å)
Form
Liquid
Quantity
100 mg
Solubility
Water
Format
Standard
Water Soluble
Yes
Product Type
Crosslinker

Contents & Storage

Upon receipt store at -20°C protected from moisture.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text