Thermo Scientific™

MT(PEG)4 Methyl-PEG-Thiol Compound

Catalog number: 26132
Have Questions?
Thermo Scientific™

MT(PEG)4 Methyl-PEG-Thiol Compound

Catalog number: 26132
Have Questions?
Catalog Number
26132
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
26132100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce MT(PEG)4, or methyl-PEG4-thiol, is a methyl- and sulfhydryl-terminated compound that contains a 4-unit polyethylene glycol (PEG) spacer and is used to modify surfaces such as quantum dots, monolayers and magnetic particles.

Features of MT(PEG)4:

Metal-binding—terminal monodentate thiol reacts spontaneously with gold surfaces through dative binding
Methyl—terminal methyl group is unreactive, the primary function of this compound being to coat quantum dot and other surfaces to reduce nonspecific binding in assays
Polyethylene glycol—PEG groups are flexible, non-immunogenic, hydrophilic, and significantly reduce nonspecific binding of surfaces for protein methods
Spacer arm—four-unit PEG spacer makes compound nearly 16 angstroms long

MT(PEG)4 is monodentate thiol-terminated and methyl pegylation reagent. The PEG compound has a defined molecular weight and spacer length and is useful for modifying surfaces such quantum dots, self-assembled monolayers and magnetic particles. Functionalization of solid surfaces with polyethylene glycol spacers such as this one significantly reduces nonspecific protein binding.

The use of MT(PEG)4 with CT(PEG)12 in surface modification can form a hydrophilic 'lawn' of methyl ether-terminated PEGs with periodic exposed carboxylic acid-containing PEGs. The exposed carboxylic acid groups can be coupled to affinity ligands using the carbodiimide coupling reaction with EDC and sulfo-NHS.

Related Products
ML(PEG)4 Methyl-PEG-Lipoamide Compound
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Metal Surface
Label Type
Polyethylene Glycol (PEG), Blocking Groups
Reactive Moiety
Carboxylic Acid, Thiol
Spacer
4 ethylene glycol units
Form
Liquid
Quantity
100 mg
Solubility
DMF or DMSO
Product Type
Thiol Compound

Contents & Storage

Upon receipt store at -20°C protected from moisture. Products shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text