Thermo Scientific™

EZ-Link™ Amine-PEG11-Biotin

Catalog number: 26136
Have Questions?
Thermo Scientific™

EZ-Link™ Amine-PEG11-Biotin

Catalog number: 26136
Have Questions?
Catalog Number
26136
Unit Size
100 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
26136100 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific EZ-Link Amine-PEG11-Biotin is a biotinylation reagent containing a long 11-unit polyethylene glycol (PEG) spacer arm and a terminal primary amine group for conjugation and protein labeling with EDC or other crosslinkers.

Features of EZ-Link Amine-PEG11-Biotin:

Biotinylation—label molecules and surfaces for assays or affinity purification methods involving avidin or streptavidin probes and resins
Amine-activated—primary amine can be crosslinked to proteins and material surfaces using EDC and other crosslinkers
Pegylated—the 11-unit, polyethylene glycol (PEG) group in spacer arm enhances water solubility of biotinylated molecules
Long reach—total reagent length is more than 50 angstroms, greatly minimizing steric hindrance for the biotin binding-function

Amine-PEG11-Biotin is the longest of three amine-modified biotin compounds that contain polyethylene glycol (PEG) spacer arms. The 11-unit PEG segment is hydrophilic and confers greater solubility to labeled proteins compared to reagents having only hydrocarbon spacers. The primary amine of this pegylated biotin reagent can be conjugated to carboxyl groups on carboxy termini, aspartate residues or glutamate residues using EDC (Part No. 22980), a water-soluble carbodiimide crosslinker. EDC activates carboxyl groups to bind to the—NH2 group of the amino-biotin, forming an amide bond.

We manufacture biotin reagents to ensure the highest possible overall product integrity, consistency and performance for the intended research applications.

Amino-biotin compounds can be conjugated to functional groups of proteins and other molecules in a variety of ways. The most common method is to crosslink the terminal primary amine to carboxyl groups using . Carboxyl groups (-COOH) occur in aspartate or glutamate residues and the carboxy-terminus of polypeptides. When activated with EDC (Part No. 22980), carboxylates react with amino (—NH2) groups to form amide bonds. Carboxylate molecules and surface materials can be pre-activated using EDC with Sulfo-NHS (Part No. 24510) for subsequent reaction to primary amines (see NHS-ester Chemistry).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Amine-PEG11-Biotin
Quantity
100 mg
Product Line
EZ-Link™
Cell Permeability
Cell-Impermeant
Chemical Reactivity
Carboxylic Acid, Ketone, Aldehyde
Reactive Moiety
Amine
Solubility
Water
Label or Dye
Biotin
Spacer
Extra-long, Pegylated
Label Type
Biotin & Analogs

Contents & Storage

Store at -20°C. Shipped at ambient temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text