Gibco™

Bovine Serum, heat inactivated, New Zealand origin

Catalog number: 26170043
Have Questions?
Gibco™

Bovine Serum, heat inactivated, New Zealand origin

Catalog number: 26170043
Have Questions?
Catalog Number
26170043
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
100 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
26170043500 mL
-
26170035100 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

Sera Category: Secure
Origin: New Zealand.
Collected from cattle typically 12-24 months of age.

Specifications

Age
12 to 24 months
Species
Cattle/Bovine
Country of Origin
New Zealand
Form
Liquid
Format
Bottle
Product Type
Adult Bovine Serum
Quantity
500 mL
Serum Treatment
Heat-inactivated
Sterility
Sterile-filtered
Green Features
Sustainable packaging
Product Line
Gibco™
Shipping Condition
Frozen

Contents & Storage

Storage conditions: ≤-10°C
Shipping conditions: Frozen

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text