Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates
Thermo Scientific™

Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates

Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityLidNo. per Pack
262162SterileWithout Lid1
262146Non-sterileWithout Lid135
268152Non-sterileWithout Lid10
268200SterileWith Lid10
Catalog number 262162
Price (USD)
254.65
Online Exclusive
270.00
Save 15.35 (6%)
Case of 50
Add to cart
Sterility:
Sterile
Lid:
Without Lid
No. per Pack:
1
Price (USD)
254.65
Online Exclusive
270.00
Save 15.35 (6%)
Case of 50
Add to cart
Improve efficiency in mixing, washing and recovery of samples with Thermo Scientific™ Nunc™ 96-Well Polystyrene Round Bottom Microwell Plates. The clear and U shaped wells are ideal for solution-based assays (e.g. agglutination assays) and common serial dilutions, allowing clean and easy pipetting. Available in a variety of options for surfaces, sterility, closures, and packaging sizes.
  • Clear polystyrene has excellent optical properties ideal for optical measurements
  • Thermo Scientific™ Nunclon™ Delta is a hydrophilic surface treatment whereas the non-treated polystyrene is more hydrophobic
  • Customized bar coding is available

Warranty: 90 days

WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceAmerican National Standards (ANSI) (ANSI/SBS 1-2004)
LidWithout Lid
No. per Case50
No. per Pack1
Packaging1 Pk.
Product TypeMicroplate
Array8 x 12
ColorClear
FormatStandard
MaterialPolystyrene
No. of Wells96
Product LineNunc
SterilitySterile
Surface TreatmentNon-treated
Well ShapeRound Bottom (U-well)
Unit SizeCase of 50

Share catalog number, name or link