Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Perform aseptic fluid transfer or sampling in biopharmaceutical applications with Thermo Scientific™ Nalgene™ Polypropylene Sanitary Carboys with 3 in. Sanitary Neck Design. The carboy's welded sanitary neck is easy to clean with no screw threads.
Autoclavable | Autoclavable |
Capacity (English) | 5.3 gal. |
Capacity (Metric) | 20 L |
Closure Size | 3 in. |
Color | White |
Description | Carboys with 3 in. Sanitary Neck Design |
Diameter (Metric) Outer | 282 mm |
For Use With (Application) | Sampling, Storage, Transportation |
Height (Metric) With Closure | 496 mm |
Includes | Handles |
Material | Polypropylene |
No. per Case | 1 Ea. |
Product Type | Carboy with Sanitary Neck |
Sterility | Non-sterile |
Unit Size | Each |
Autoclavable | Autoclavable |
Capacity (English) | 2.6 gal. |
Capacity (Metric) | 10 L |
Closure Size | 3 in. |
Color | White |
Description | Carboys with 3 in. Sanitary Neck Design |
Diameter (Metric) Outer | 250 mm |
For Use With (Application) | Sampling, Storage, Transportation |
Height (Metric) With Closure | 353 mm |
Includes | Handles |
Material | Polypropylene |
No. per Case | 1 Ea. |
Product Type | Carboy with Sanitary Neck |
Sterility | Non-sterile |
Unit Size | Each |
Autoclavable | Autoclavable |
Capacity (English) | 13 gal. |
Capacity (Metric) | 50 L |
Closure Size | 3 in. |
Color | White |
Description | Carboys with 3 in. Sanitary Neck Design |
Diameter (Metric) Outer | 379 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Sampling, Storage |
Height (Metric) With Closure | 545 mm |
Includes | Handles |
Material | Polypropylene |
No. per Case | 1 Ea. |
Product Type | Carboy with Sanitary Neck |
Sterility | Non-sterile |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Capacity (Metric) | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2630-0020 | Each | 20 L | 523.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2630-0010 | Each | 10 L | 401.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2630-0050 | Each | 50 L | 668.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.