Thermo Scientific Chemicals

Tetrahydro-2-furoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 263145000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetrahydro-2-furoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 263145000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 263145000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16874-33-2
IUPAC Name
oxolane-2-carboxylic acid
Molecular Formula
C5H8O3
InChI Key
UJJLJRQIPMGXEZ-UHFFFAOYNA-N
SMILES
OC(=O)C1CCCO1
Titration with NaOH
98.0 to 102.0 %
Appearance (Form)
Liquid
Refractive index
1.4580 to 1.4620 (20°C, 589 nm)
GC
>=98.0 %
Appearance (Color)
Clear colorless to yellow to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text