Thermo Scientific Chemicals

Butyric acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 263190250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Butyric acid sodium salt, 98%, Thermo Scientific Chemicals

Catalog number: 263190250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 263190250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
156-54-7
IUPAC Name
sodium butanoate
Molecular Formula
C4H7NaO2
InChI Key
MFBOGIVSZKQAPD-UHFFFAOYSA-M
SMILES
[Na+].CCCC([O-])=O
Appearance (Color)
White
Infrared spectrum
Conforms
Titration with HClO4
97.5 to 102.5 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text