2,5-Dibromotoluene, 98%, Thermo Scientific Chemicals
2,5-Dibromotoluene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,5-Dibromotoluene, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Packaging:
Glass bottle
Catalog number 263480250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS615-59-8
IUPAC Name1,4-dibromo-2-methylbenzene
Molecular FormulaC7H6Br2
InChI KeyQKEZTJYRBHOKHH-UHFFFAOYSA-N
SMILESCC1=CC(Br)=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.6000 to 1.6040 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only