Thermo Scientific Chemicals

Cyclopropylamine, 98%, Thermo Scientific Chemicals

Catalog number: 263680025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

Cyclopropylamine, 98%, Thermo Scientific Chemicals

Catalog number: 263680025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 263680025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
765-30-0
IUPAC Name
cyclopropanamine
Molecular Formula
C3H7N
InChI Key
HTJDQJBWANPRPF-UHFFFAOYSA-N
SMILES
NC1CC1
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.4190 to 1.4220 (20°C, 589 nm)
GC
>=97.5 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text