Cyclopropylamine, 98%, Thermo Scientific Chemicals
Cyclopropylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclopropylamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 263680250
Price (USD)/ Each
77.65
Online exclusive
86.10 
Save 8.45 (10%)
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
77.65
Online exclusive
86.10 
Save 8.45 (10%)
Add to cart
Cyclopropylamine, 98%, Thermo Scientific Chemicals
Catalog number263680250
Price (USD)/ Each
77.65
Online exclusive
86.10 
Save 8.45 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Namecyclopropanamine
Molecular FormulaC3H7N
InChI KeyHTJDQJBWANPRPF-UHFFFAOYSA-N
SMILESNC1CC1
Molecular Weight (g/mol)57.10
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4190 to 1.4220 (20°C, 589 nm)
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only