Zinc stearate
Zinc stearate
Zinc stearate
Thermo Scientific Chemicals

Zinc stearate

CAS: 557-05-1 | C36H70O4Zn | 632.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
20 kg
1 kg
5 kg
Catalog number 263780010
Price (EUR)
28,82
Online Exclusive
33,90
Save 5,08 (15%)
Each
-
Add to cart
Quantity:
1 kg
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
28,82
Online Exclusive
33,90
Save 5,08 (15%)
Each
Add to cart
Chemical Identifiers
CAS557-05-1
IUPAC Namezinc(2+) dioctadecanoate
Molecular FormulaC36H70O4Zn
InChI KeyXOOUIPVCVHRTMJ-UHFFFAOYSA-L
SMILES[Zn++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Complexometric12.5 to 14.0 % (ZnO)
Infrared spectrumConforms
Appearance (Color)White
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472324Certificate of AnalysisMay 17, 2025263780200, 263780050, 263780010
A0472521Certificate of AnalysisMay 17, 2025263780200, 263780050, 263780010
A0465378Certificate of AnalysisAug 15, 2024263780200, 263780050, 263780010
A0462025Certificate of AnalysisApr 22, 2024263780200, 263780050, 263780010
A0397789Certificate of AnalysisDec 14, 2022263780200, 263780050, 263780010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

EU Specific Hazard Statements

May form combustible dust concentrations in air

Share catalog number, name or link

1x1 image pixel for data collection