Potassium phosphate, tribasic monohydrate, 96%, extra pure, Thermo Scientific Chemicals
Potassium phosphate, tribasic monohydrate, 96%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium phosphate, tribasic monohydrate, 96%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
25 g
500 g
1 kg
25 kg
Packaging:
View all
Plastic Bottle
Catalog number 263790050
Price (USD)/ Each
-
Quantity:
5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS27176-10-9
IUPAC Nametripotassium hydrate phosphate
Molecular FormulaH2K3O5P
InChI KeyRMNIZOOYFMNEJJ-UHFFFAOYSA-K
SMILESO.[K+].[K+].[K+].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or granules
Loss on ignition=<13 % (800°C)
Iron (Fe)=<20 ppm
Appearance (Color)White
Titration excess HCl>=95 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only