Magnesium oxide, 98%, extra pure, powder, particle size: 99% <150 μm (-100 mesh)
Magnesium oxide, 98%, extra pure, powder, particle size: 99% &lt;150 &mu;m (-100 mesh)
Magnesium oxide, 98%, extra pure, powder, particle size: 99% &lt;150 &mu;m (-100 mesh)
Magnesium oxide, 98%, extra pure, powder, particle size: 99% &lt;150 &mu;m (-100 mesh)
Magnesium oxide, 98%, extra pure, powder, particle size: 99% &lt;150 &mu;m (-100 mesh)
Thermo Scientific Chemicals

Magnesium oxide, 98%, extra pure, powder, particle size: 99% <150 μm (-100 mesh)

CAS: 1309-48-4 | MgO | 40.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 263835000
Price (USD)
105.00
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
105.00
Each
Add to cart
Chemical Identifiers
CAS1309-48-4
IUPAC Nameoxomagnesium
Molecular FormulaMgO
InChI KeyCPLXHLVBOLITMK-UHFFFAOYSA-N
SMILESO=[Mg]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle size=<1 % Residue on 150 µm (>100 mesh) sieve
Ask our AI about this Product

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473027Certificate of AnalysisMay 29, 2025263830025, 263830100, 263835000
A0473028Certificate of AnalysisMay 28, 2025263830025, 263830100, 263835000
A0469418Certificate of AnalysisFeb 05, 2025263830025, 263830100, 263835000
A0469345Certificate of AnalysisJan 31, 2025263830025, 263830100, 263835000
A0467997Certificate of AnalysisJan 31, 2025263830025, 263830100, 263835000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection