Nalgene™ Polypropylene, Carboy with Sanitary Flange
Thermo Scientific™

Nalgene™ Polypropylene, Carboy with Sanitary Flange

For Use With: Supply reservoirs to systems such as fermentors or chromatography columns.; Autoclavable; Closure Material: Polypropylene; Color: White
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
2640-002010 L
2640-005020 L
2640-013050 L
Catalog number 2640-0020
Price (USD)
806.00
Each
Capacity (Metric):
10 L
Price (USD)
806.00
Each

Use the Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Carboys with 1.5 in. Sanitary Flanges as dispensing ports or with secure hookup for supply reservoirs to systems such as fermentors or chromatography columns.

Key Features:

  • Molded-in handles for ease in transporting containers
  • Molded-in graduations in 1 gal. and 5 L increments
  • USP VI, autoclavable, graduated, leakproof†

Includes:

Size 83B closure; accepts No. 13.5 rubber stopper

Note:

Follow autoclave instructions provided with the product for proper function

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity (English)2.6 gal.
Certifications/ComplianceUSP Class VI, non-cytotoxic
Closure MaterialPolypropylene
Closure Size83B
Diameter (Metric) Outer250 mm
For Use With (Application)Sampling, Storage, Transportation ​
GraduationsGraduated to contain in 5 L/1 gal. increments
IncludesMolded-in handles, 1.5 in. secure, non-threaded sanitary dispensing port
Volume (English) Brim3 gal.
Volume (Metric) Brim12 L
Workflow StepUpstream media/buffer prep and storage
AutoclavableAutoclavable
Capacity (Metric)10 L
ColorWhite
MaterialPolypropylene
Product TypeCarboy with Sanitary Flange
SterilityNon-sterile
Unit SizeEach