Thermo Scientific Chemicals

N,N-Dimethylacetamide dimethyl acetal, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 264130250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N-Dimethylacetamide dimethyl acetal, 97%, stabilized, Thermo Scientific Chemicals

Catalog number: 264130250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 264130250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
18871-66-4
IUPAC Name
(1,1-dimethoxyethyl)dimethylazanium
Molecular Formula
C6H16NO2
InChI Key
FBZVZUSVGKOWHG-UHFFFAOYSA-O
SMILES
COC(C)(OC)[NH+](C)C
Appearance (Form)
Liquid
Refractive index
1.4100 to 1.4180 (20°C, 589 nm)
Stabilizer
sodium carbonate
Titration with HClO4
>=96.0 %
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text