Thermo Scientific Chemicals

Methyl Red, pure, indicator

Catalog number: 264440250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl Red, pure, indicator

Catalog number: 264440250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 264440250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
493-52-7
IUPAC Name
2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid
Molecular Formula
C15H15N3O2
InChI Key
CEQFOVLGLXCDCX-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O
Appearance (Color)
Dark red to brown to purple to violet
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Solubility
(0.1 % in ethanol) Clear
Visual transition interval
from pH 4.2 (pink or red) to pH 6.2 (yellow)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text