3-Amino-1H-1,2,4-triazole, 95%, Thermo Scientific Chemicals
3-Amino-1H-1,2,4-triazole, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-1H-1,2,4-triazole, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 264570100
Price (USD)/ Each
27.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
31.90 
Save 4.25 (13%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
27.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
31.90 
Save 4.25 (13%)
Add to cart
3-Amino-1H-1,2,4-triazole, 95%, Thermo Scientific Chemicals
Catalog number264570100
Price (USD)/ Each
27.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
31.90 
Save 4.25 (13%)
-
Add to cart
Chemical Identifiers
CAS61-82-5
IUPAC Name1H-1,2,4-triazol-5-amine
Molecular FormulaC2H4N4
InChI KeyKLSJWNVTNUYHDU-UHFFFAOYSA-N
SMILESNC1=NC=NN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or flakes
Titration with HClO4>=94 %
Appearance (Color)White to off-white to yellow
Infrared spectrumConforms
Melting point147°C to 156°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only