Thermo Scientific Chemicals

(2S)-(+)-Glycidyl tosylate, 99+%, Thermo Scientific Chemicals

Catalog number: 264580050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

(2S)-(+)-Glycidyl tosylate, 99+%, Thermo Scientific Chemicals

Catalog number: 264580050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 264580050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
70987-78-9
IUPAC Name
[(2S)-oxiran-2-yl]methyl 4-methylbenzene-1-sulfonate
Molecular Formula
C10H12O4S
InChI Key
NOQXXYIGRPAZJC-VIFPVBQESA-N
SMILES
CC1=CC=C(C=C1)S(=O)(=O)OC[C@@H]1CO1
Appearance (Form)
Powder
GC
>=99.0 %
Infrared spectrum
Conforms
Appearance (Color)
White
Melting point
45°C to 48°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text