(2S)-(+)-Glycidyl tosylate, 99+%, Thermo Scientific Chemicals
(2S)-(+)-Glycidyl tosylate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(2S)-(+)-Glycidyl tosylate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 264580250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS70987-78-9
IUPAC Name[(2S)-oxiran-2-yl]methyl 4-methylbenzene-1-sulfonate
Molecular FormulaC10H12O4S
InChI KeyNOQXXYIGRPAZJC-VIFPVBQESA-N
SMILESCC1=CC=C(C=C1)S(=O)(=O)OC[C@@H]1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
GC>=99.0 %
Infrared spectrumConforms
Appearance (Color)White
Melting point45°C to 48°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only