Thermo Scientific Chemicals

(S)-(+)-2-Phenylpropionic acid, 99%, Thermo Scientific Chemicals

Catalog number: 264652500
250 mg, Glass Bottle, Each
Thermo Scientific Chemicals

(S)-(+)-2-Phenylpropionic acid, 99%, Thermo Scientific Chemicals

Catalog number: 264652500
250 mg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 264652500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7782-24-3
IUPAC Name
2-phenylpropanoic acid
Molecular Formula
C9H10O2
InChI Key
YPGCWEMNNLXISK-UHFFFAOYNA-N
SMILES
CC(C(O)=O)C1=CC=CC=C1
Appearance (Form)
Low melting solid
Melting point
29°C to 32°C
Specific optical rotation
+74° to +76° (20°C, 589 nm) (c=3, CHCl3)
Infrared spectrum
Conforms
Appearance (Color)
Off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text