Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 kg
Packaging:
View all
Glass bottle
Catalog number 264755000
Price (USD)/ Each
56.65
Online exclusive
65.90 
Save 9.25 (14%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
56.65
Online exclusive
65.90 
Save 9.25 (14%)
Add to cart
Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals
Catalog number264755000
Price (USD)/ Each
56.65
Online exclusive
65.90 
Save 9.25 (14%)
-
Add to cart
Chemical Identifiers
IUPAC NameMolecular sieves
Synonym2-[2-acetamido-3-(3-fluorophenyl)propanamido]-5-methoxy-4-(methoxycarbonyl)-3-methyl-5-oxopentanoic acid
MDL NumberMFCD00131613
CAS70955-01-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Granules
Water adsorption>=20 % (50% rH, 20°C, 24 h)
Bulk density680 to 780 g/l
Appearance (Color)Beige
Residual water=<1.5 % (50°C, 2 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
RUO – Research Use Only