Hypersil GOLD™ PEI HILIC HPLC Columns
Hypersil GOLD™ PEI HILIC HPLC Columns
Actual product may vary
Thermo Scientific™

Hypersil GOLD™ PEI HILIC HPLC Columns

Retain highly polar and hydrophilic analytes, which are difficult to analyze with conventional C18 columns, using Hypersil GOLD PEI HILIC LC Columns.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberParticle SizeDiameter (Metric)Length (Metric)
26502-1021301.9 μm2.1 mm100 mm
26505-0521305 μm2.1 mm50 mm
26505-1021305 μm2.1 mm100 mm
26505-1521305 μm2.1 mm150 mm
26505-1546305 μm4.6 mm150 mm
26505-2546305 μm4.6 mm250 mm
26502-1521301.9 μm2.1 mm150 mm
26502-0521301.9 μm2.1 mm50 mm
26503-1546303 μm4.6 mm150 mm
26503-1530303 μm3 mm150 mm
26503-1521303 μm2.1 mm150 mm
26503-1510303 μm1 mm150 mm
26503-1030303 μm3 mm100 mm
26503-1021303 μm2.1 mm100 mm
26503-0521303 μm2.1 mm50 mm
26505-1030305 μm3 mm100 mm
26503-0121013 μm2.1 mm10 mm
26503-0110013 μm1 mm10 mm
26505-0140015 μm4 mm10 mm
19 Options
Catalog number 26502-102130
Price (USD)/ Each
1,096.34
Add to cart
Particle Size:
1.9 μm
Diameter (Metric):
2.1 mm
Length (Metric):
100 mm
Price (USD)/ Each
1,096.34
Add to cart
Hypersil GOLD™ PEI HILIC HPLC Columns
Catalog number26502-102130
Price (USD)/ Each
1,096.34
-
Add to cart
Retain highly polar and hydrophilic analytes, which are difficult to analyze with conventional C18 columns, using Thermo Scientific™ Hypersil GOLD™ PEI HILIC (hydrophilic interaction liquid chromatography) LC Columns. These columns deliver straightforward separation without the need for sample derivatization or ion-pair reagents. Featuring outstanding peak shape and selectivity, the columns provide improved sensitivity for MS detection. With 1.9, 3 and 5μm particle sizes, Hypersil GOLD PEI HILIC Columns are ideal for applications within drug discovery, food testing and environmental analysis.
About the Hypersil GOLD Family of HPLC Columns:

Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3 and 5μm particle sizes for all your separation needs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHILIC, ESI-MS
Length (Metric)100 mm
Max. Pressure18130 psi (1250 bar)
Particle Size1.9 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit SizeEach