2-Tienilmercaptano, 98 %, Thermo Scientific Chemicals
2-Tienilmercaptano, 98 %, Thermo Scientific Chemicals
2-Tienilmercaptano, 98 %, Thermo Scientific Chemicals
2-Tienilmercaptano, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Tienilmercaptano, 98 %, Thermo Scientific Chemicals

CAS: 6258-63-5 | C5H6S2 | 130.22 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Envase:
Frasco de vidrio
Número de catálogo 265110010
Precio (EUR)
140,25
Precio exclusivo en nuestra web
165,00
Ahorro 24,75 (15%)
Each
-
Añadir al carro de la compra
Cantidad:
1 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
140,25
Precio exclusivo en nuestra web
165,00
Ahorro 24,75 (15%)
Each
Añadir al carro de la compra
Identificadores químicos
CAS6258-63-5
IUPAC Name(thiophen-2-yl)methanethiol
Molecular FormulaC5H6S2
InChI KeyGCZQHDFWKVMZOE-UHFFFAOYSA-N
SMILESSCC1=CC=CS1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.6020 to 1.6040 (20°C, 589 nm)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documentos y descargas

Certificados

N.º de loteCertificate TypeDateCatalog Number(s)
A0468505Certificate of Analysis17 may 2025265110050, 265110010
A0410927Certificate of Analysis13 dic 2022265110050, 265110010
A0415458Certificate of Analysis18 ago 2022265110050, 265110010
Se muestran 3 resultados, busque arriba un certificado específico

Hojas de datos de seguridad

Seguridad y manipulación

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Label Elements
Signal Word

Warning

Hazard Statements

H302 - Harmful if swallowed

EU Specific Hazard Statements

Combustible liquid

Precautionary Statements

P264 - Wash face, hands and any exposed skin thoroughly after handling

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P312 - Call a POISON CENTER or doctor if you feel unwell

Compartir número de catálogo, nombre o enlace.

1x1 image pixel for data collection