Search by lot number or partial lot number
Thermo Scientific™ Nunc 384-Well Clear Polystyrene Plates with Cell Culture Treated and Nontreated Surfaces are optimized for robotics, plate readers and liquid handling systems. 384-well microplates allow sample and reagent volume reduction providing cost savings and allowing more samples to be screened. Our plates are high quality for consistent and reliable results.
Barcode | Custom Barcoding Available |
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Lid | Without Lid |
Culture Area | 0.084 cm2/well |
Description | Non-Treated; no lid; Sterile; 25/100 |
Array | 16 x 24 |
Packaging | 25/Pk., 100/Cs. |
Product Line | Nunc |
Color | Clear |
Product Type | Microplate |
Material | Polystyrene |
Volume (Metric) Well | 120 μL |
No. of Wells | 384 |
Volume (Metric) Working | 10 to 100 μL |
Sterility | Sterile |
Surface Treatment | Non-treated |
Well Shape | Flat Bottom (F-well) |
No. per Case | 100 |
No. per Pack | 25 |
Unit Size | Case of 100 |
Catalog Number | Specifications | Unit Size | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
265203 | Case of 100 | 764.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|