2,2,4-Trimethylpentane, 99.5%, for analysis, Thermo Scientific Chemicals
2,2,4-Trimethylpentane, 99.5%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,4-Trimethylpentane, 99.5%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 265440010
Price (USD)/ Each
75.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
88.40 
Save 12.75 (14%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
75.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
88.40 
Save 12.75 (14%)
Add to cart
2,2,4-Trimethylpentane, 99.5%, for analysis, Thermo Scientific Chemicals
Catalog number265440010
Price (USD)/ Each
75.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
88.40 
Save 12.75 (14%)
-
Add to cart
Chemical Identifiers
CAS540-84-1
IUPAC Name2,2,4-trimethylpentane
Molecular FormulaC8H18
InChI KeyNHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILESCC(C)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Water=<0.01 % (Coulometric)
Nickel (Ni)=<0.1 ppm
Aluminium (Al)=<0.5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only