Thermo Scientific Chemicals

Thallium(I) nitrate, 99.5%, Thermo Scientific Chemicals

Catalog number: 265590250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Thallium(I) nitrate, 99.5%, Thermo Scientific Chemicals

Catalog number: 265590250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 265590250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10102-45-1
IUPAC Name
λ¹-thallanylium nitrate
Molecular Formula
NO3Tl
InChI Key
FYWSTUCDSVYLPV-UHFFFAOYSA-N
SMILES
[Tl+].[O-][N+]([O-])=O
Appearance (Color)
White
Total trace metal impurities
=<5000 ppm
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text