Thermo Scientific Chemicals

Ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: 265640010
1 g, Glass Bottle, Each
Thermo Scientific Chemicals

Ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: 265640010
1 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 265640010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1127-99-7
IUPAC Name
hydrogen ethyl (1R,2S)-2-aminocyclohexane-1-carboxylate chloride
Molecular Formula
C9H18ClNO2
InChI Key
XMQSOBPCWYVZSW-WLYNEOFISA-N
SMILES
[H+].[Cl-].CCOC(=O)[C@@H]1CCCC[C@@H]1N
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder and/or chunks
Melting point
128°C to 135°C
Titration Argentometric
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text