Thermo Scientific Chemicals

3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 265690010
Each
Thermo Scientific Chemicals

3-exo-Aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 265690010
Each
Catalog number: 265690010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
92511-32-5
IUPAC Name
3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylic acid
Molecular Formula
C8H11NO2
InChI Key
FCYFJGGJCJDCPB-UHFFFAOYNA-N
SMILES
NC1C2CC(C=C2)C1C(O)=O
Appearance (Form)
Powder
Water
=<10.5 % (K.F.)
Titration with HClO4
>=97.5 % (On dry substance)
Infrared spectrum
Conforms
Appearance (Color)
Beige to light grey

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text