Thermo Scientific Chemicals

cis-2-Aminocyclohexanol hydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: 265850050
5 g, Each
Thermo Scientific Chemicals

cis-2-Aminocyclohexanol hydrochloride, 99+%, Thermo Scientific Chemicals

Catalog number: 265850050
5 g, Each
Quantity
Catalog number: 265850050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6936-47-6
IUPAC Name
(1S,2R)-2-aminocyclohexan-1-ol hydrochloridyl
Molecular Formula
C6H13ClNO
InChI Key
SEBSRZIHRZRNAH-IBTYICNHSA-N
SMILES
[Cl].N[C@@H]1CCCC[C@@H]1O
Appearance (Form)
Crystalline powder
Melting point
186°C to 190°C
Infrared spectrum
Conforms
Appearance (Color)
White to light beige
Titration with HClO4
94 to 106 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text