Thermo Scientific Chemicals

cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 265860010
Each
Thermo Scientific Chemicals

cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 265860010
Each
Catalog number: 265860010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5691-37-2
IUPAC Name
(1S,2R)-2-(hydroxymethyl)cyclohexan-1-aminium
Molecular Formula
C7H16NO
InChI Key
GCWPGEWXYDEQAY-BQBZGAKWSA-O
SMILES
[NH3+][C@H]1CCCC[C@H]1CO
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
Titration Argentometric
>=98.5 %
Appearance (Form)
Crystalline powder
Melting point
131°C to 135°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text