Thermo Scientific Chemicals

1-Methyl-6,7-dimethoxy-3,4-dihydroisoquinoline, 99+%, Thermo Scientific Chemicals

Catalog number: 265970010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Methyl-6,7-dimethoxy-3,4-dihydroisoquinoline, 99+%, Thermo Scientific Chemicals

Catalog number: 265970010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 265970010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4721-98-6
IUPAC Name
6,7-dimethoxy-1-methyl-3,4-dihydroisoquinoline
Molecular Formula
C12H15NO2
InChI Key
VASUQTGZAPZKFK-UHFFFAOYSA-N
SMILES
COC1=CC2=C(C=C1OC)C(C)=NCC2
Appearance (Color)
Light yellow to beige
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder
GC
>=99.0 %
Melting point
103°C to 107°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text