PageRuler™ Plus Prestained Protein Ladder, 10 to 250 kDa
PageRuler™ Plus Prestained Protein Ladder, 10 to 250 kDa
Thermo Scientific™

PageRuler™ Plus Prestained Protein Ladder, 10 to 250 kDa

Thermo Scientific PageRuler Plus Prestained Protein Ladder is a mixture of nine blue-, orange-, and green-stained proteins (10 to 250Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
266192 x 250 μL
2662010 x 250 μL
Catalog number 26619
Price (USD)
171.65
Online Exclusive
202.00
Save 30.35 (15%)
Each
-
Add to cart
Quantity:
2 x 250 μL
Request bulk or custom format
Price (USD)
171.65
Online Exclusive
202.00
Save 30.35 (15%)
Each
Add to cart
Ask our AI about this Product

Thermo Scientific PageRuler Plus Prestained Protein Ladder is a mixture of nine blue-, orange-, and green-stained proteins (10 to 250 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Applications

  • Monitoring protein migration during SDS-polyacrylamide gel electrophoresis
  • Monitoring protein transfer onto membranes after western blotting
  • Sizing of proteins on SDS-polyacrylamide gels and western blots
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodColorimetric, NIR Fluorescence (700 nm), RGB Fluorescence (555 nm)
Gel CompatibilityBolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Molecular Weight (g/mol)250, 130, 100, 70, 55, 35, 25, 15, 10 kDa
Product LinePageRuler™
Product TypeProtein Ladder
Quantity2 x 250 μL
Ready to LoadYes
Shipping ConditionApproved for shipment on Wet or Dry Ice
Stain Type3 colors: Blue, Orange, Green
System TypeWestern Blotting, SDS-PAGE
Number of Markers9
Size Range10 to 250 kDa
Unit SizeEach
Contents & Storage

Contents: two vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol

Storage: Upon receipt store at -20°C

iBlot 3 Western Blot Transfer System

Introducing iBlot 3 Western Blot Transfer System

Featuring higher throughput and built-in cooling for consistent protein transfer
Learn more

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link