Search Thermo Fisher Scientific
Use Thermo Scientific™ Nalgene™ PVDF True Union Clamps with silicone gasket and end caps to experience reliable, repeatable seals. Molded of PVDF fluoride, these threaded clamps are a fully autoclavable clamp system.
Quantity | 1/Pk., 1/Cs. |
Type | Mini |
Material | PVDF |
For Use With (Equipment) | Nalgene™ Sanitary Carboys, Nalgene™ Autoclavable Carboys with Spigot |
Size (English) Grip | 0.75 in. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
2670-0075 | Each | 53.25 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|